Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039350.2 | NP_001034439.1 | 14 | 1868 | ||
NM_001290252.1 | NP_001277181.1 | 12 | 1435 | |||
NM_001936.4 | NP_001927.3 | 14 | 1570 | |||
NM_130797.3 | NP_570629.2 | 14 | 1876 | |||
XM_017011808.1 | XP_016867297.1 | 15 | 1822 | |||
XM_017011809.1 | XP_016867298.1 | 15 | 1585 | |||
XM_017011810.1 | XP_016867299.1 | 15 | 1688 | |||
XM_017011811.1 | XP_016867300.1 | 15 | 1612 | |||
XM_017011812.1 | XP_016867301.1 | 10 | 983 | |||
GenBank mRNA | AK293582.1 | - | 7 | 747 | ||
AK294483.1 | - | 12 | 1205 | |||
BC150304.1 | - | 14 | 1867 | 8860401 | ||
BX647966.1 | - | 14 | 1633 | |||
DA911296.1 | - | 2 | 129 | |||
M96859.1 | - | 14 | 1617 | |||
M96860.1 | - | 14 | 1464 |