Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001385.2 | NP_001376.1 | 8 | 1512 | ||
XM_005250818.3 | XP_005250875.1 | 9 | 1645 | |||
XM_006716518.3 | XP_006716581.1 | 8 | 1486 | |||
XM_011516903.2 | XP_011515205.1 | 9 | 1646 | |||
GenBank mRNA | AK311261.1 | - | 2 | 287 | ||
BC034395.1 | - | 8 | 1476 | 5187250 | ||
D78011.1 | - | 8 | 1512 |