Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000795.3 | NP_000786.1 | 4, 5 | 774 | ||
NM_016574.3 | NP_057658.2 | 4, 5 | 774 | |||
XM_017017296.1 | XP_016872785.1 | 4, 5 | 756 | |||
GenBank mRNA | AB209832.1 | - | 3, 4 | 569 | ||
AB451480.1 | - | 3, 4 | 539 | |||
AF176812.1 | - | 3, 4 | 548 | |||
AK308777.1 | - | 4, 5 | 604 | |||
BC021195.2 | - | 4, 5 | 634 | 3939741 | ||
M29066.1 | - | 3, 4 | 572 | |||
M30625.1 | - | 4, 5 | 875 | |||
S62137.1 | - | 3, 4 | 856 | |||
S69899.1 | - | 4, 5 | 638 | |||
X51362.1 | - | 4, 5 | 704 | |||
X51645.1 | - | 3, 4 | 567 |
Set Membership: |
G-protein coupled receptor |