Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
siRNA ID
HSS102972
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
4
454
NM_001256304.1
NP_001243233.1
3
191
NM_001256308.2
NP_001243237.1
3
325
NM_001320932.1
NP_001307861.1
4
454
NM_001320933.1
NP_001307862.1
4
454
NM_001320934.1
NP_001307863.1
4
454
NM_001320935.1
NP_001307864.1
4
454
NM_001320936.1
NP_001307865.1
4
454
NM_001320937.1
NP_001307866.1
4
454
NM_021907.4
NP_068707.1
4
454
NM_033147.3
NP_149159.2
4
454
NM_033148.3
NP_149160.1
4
454
NM_183360.2
NP_899204.1
4
454
NM_183361.2
NP_899205.1
4
454
XM_005264182.1
XP_005264239.1
4
371
XM_011532671.1
XP_011530973.1
4
371
XM_011532672.2
XP_011530974.1
4
371
XM_011532673.1
XP_011530975.1
4
371
XM_011532674.1
XP_011530976.1
4
371
XM_011532675.1
XP_011530977.1
4
371
XM_011532676.1
XP_011530978.1
4
371
XM_011532677.2
XP_011530979.1
3
827
XM_011532679.1
XP_011530981.1
4
371
XM_011532680.2
XP_011530982.1
4
378
XM_011532681.2
XP_011530983.1
4
371
XM_017003529.1
XP_016859018.1
4
371
XM_017003530.1
XP_016859019.1
4
371
XM_017003531.1
XP_016859020.1
4
371
XM_017003532.1
XP_016859021.1
4
371
XM_017003533.1
XP_016859022.1
4
371
XM_017003534.1
XP_016859023.1
4
371
XM_017003535.1
XP_016859024.1
4
371
XM_017003536.1
XP_016859025.1
4
371
XM_017003538.1
XP_016859027.1
4
303
XM_017003539.1
XP_016859028.1
4
314
XM_017003540.1
XP_016859029.1
4
371
XM_017003541.1
XP_016859030.1
4
355
XM_017003542.1
XP_016859031.1
4
378
XM_017003543.1
XP_016859032.1
4
358
XM_017003544.1
XP_016859033.1
4
378
XM_017003545.1
XP_016859034.1
4
358
GenBank mRNA
AF022728.1
-
4
352
AK225724.1
-
4
357
AK294174.1
-
4
359
AK294357.1
-
4
221
AK300019.1
-
4
331
AK301386.1
-
3
327
AL555818.3
-
4
368
BC016655.1
-
4
347
3868207
BC049366.1
-
4
445
4817552
BT009805.1
-
3
191
BU846095.1
-
2
132
DB460644.1
-
4
454
DQ160290.1
-
3
191
Y12712.1
-
4
374
Y15718.1
-
3
191
Y15722.1
-
3
191
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline