Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004417.3 | NP_004408.1 | 1, 2 | 618 | ||
GenBank mRNA | AK298047.1 | - | 2, 3 | 552 | ||
AK298658.1 | - | 2, 3 | 418 | |||
AK299391.1 | - | 2, 3 | 622 | |||
AK300487.1 | - | 2, 3 | 532 | |||
AK304475.1 | - | 2, 3 | 410 | |||
BC022463.1 | - | 1, 2 | 613 | 4794895 | ||
X68277.1 | - | 1, 2 | 603 |