Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000113.2 | NP_000104.1 | 2 | 356 | ||
GenBank mRNA | AF007871.1 | - | 2 | 321 | ||
AK056062.1 | - | 2 | 328 | |||
AK075343.1 | - | 2 | 306 | |||
AK294677.1 | - | 2 | 446 | |||
AK314505.1 | - | 2 | 300 | |||
BC000674.2 | - | 2 | 317 | 3347611 | ||
BC014484.1 | - | 2 | 298 | 4869856 | ||
BT006931.1 | - | 2 | 279 |