Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EBF1 SIRNA Assays ›
Gene Symbol:
EBF1
Gene Name:
early B-cell factor 1
Gene Aliases:
COE1, EBF, O/E-1, OLF1
Chromosome Location:
Chr.5: 158695915 - 159099762 on Build GRCh38
UniGene ID:
Hs.573143
Species:
Human
siRNA ID
HSS103036
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EBF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001290360.2
NP_001277289.1
13
1599
NM_001324101.1
NP_001311030.1
13
1599
NM_001324103.1
NP_001311032.1
13
1596
NM_001324106.1
NP_001311035.1
13
1572
NM_001324107.1
NP_001311036.1
13
1575
NM_001324108.1
NP_001311037.1
13
1572
NM_001324109.1
NP_001311038.1
13
1599
NM_001324111.1
NP_001311040.1
13
1694
NM_024007.4
NP_076870.1
13
1596
NM_182708.2
NP_874367.1
12
1503
XM_017009192.1
XP_016864681.1
13
1599
XM_017009193.1
XP_016864682.1
13
1584
XM_017009194.1
XP_016864683.1
13
1575
XM_017009195.1
XP_016864684.1
13
1572
XM_017009196.1
XP_016864685.1
13
2534
XM_017009197.1
XP_016864686.1
13
2037
XM_017009198.1
XP_016864687.1
13
1956
XM_017009199.1
XP_016864688.1
8
1950
XM_017009200.1
XP_016864689.1
8
19086
XM_017009201.1
XP_016864690.1
8
19083
XM_017009204.1
XP_016864693.1
5
1476
GenBank mRNA
AF208502.1
-
13
1298
AK289712.1
-
13
1583
AK303698.1
-
11
1225
AK304438.1
-
13
1302
BC038805.1
-
13
1607
4524661
BC041178.1
-
12
1430
6045572
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
multicellular organism development
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
metal ion binding
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline