Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001406.3 | NP_001397.1 | 2 | 535 | ||
GenBank mRNA | AK314841.1 | - | 2 | 641 | ||
BC042944.2 | - | 2 | 535 | 5286243 | ||
U57001.1 | - | 2 | 535 | |||
U62775.1 | - | 2 | 339 | |||
U66406.1 | - | 2 | 751 |