Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178130.2 | NP_001171601.1 | 21, 22 | 3617 | ||
NM_001178131.2 | NP_001171602.1 | 21, 22 | 3614 | |||
NM_001963.5 | NP_001954.2 | 22, 23 | 3740 | |||
XM_005262796.2 | XP_005262853.1 | 22, 23 | 3741 | |||
XM_011531707.1 | XP_011530009.1 | 23, 24 | 3390 | |||
XM_017007845.1 | XP_016863334.1 | 22, 23 | 3765 | |||
XM_017007846.1 | XP_016863335.1 | 22, 23 | 3765 | |||
XM_017007847.1 | XP_016863336.1 | 21, 22 | 3642 | |||
XM_017007848.1 | XP_016863337.1 | 21, 22 | 3639 | |||
XM_017007849.1 | XP_016863338.1 | 20, 21 | 3522 | |||
GenBank mRNA | AK299306.1 | - | 21, 22 | 3616 | ||
BC093731.1 | - | 22, 23 | 3383 | 7939576 | ||
BC113461.1 | - | 22, 23 | 3383 | 8322513 | ||
BC143357.1 | - | 21, 22 | 3261 | |||
X04571.1 | - | 22, 23 | 3724 |