Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199880.1 | NP_001186809.1 | 2 | 509 | ||
NM_001199881.1 | NP_001186810.1 | 2 | 503 | |||
NM_004430.2 | NP_004421.2 | 2 | 933 | |||
XM_005273425.3 | XP_005273482.1 | 2 | 884 | |||
XM_005273426.3 | XP_005273483.1 | 3 | 557 | |||
XM_011544429.2 | XP_011542731.1 | 3 | 669 | |||
GenBank mRNA | AK292464.1 | - | 2 | 829 | ||
AK294968.1 | - | 2 | 468 | |||
AK295134.1 | - | 2 | 510 | |||
AK313604.1 | - | 2 | 829 | |||
BC104765.1 | - | 2 | 674 | 8143768 | ||
BC112279.1 | - | 2 | 672 | 8327747 | ||
S40832.1 | - | 2 | 932 | |||
X63741.1 | - | 2 | 928 |