Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4EBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004096.4 | NP_004087.1 | 1, 2 | 452 | ||
GenBank mRNA | AK313724.1 | - | 1, 2 | 412 | ||
BC005057.1 | - | 1, 2 | 178 | 2823322 | ||
BC050633.1 | - | 1, 2 | 391 | 5555034 | ||
BG717474.1 | - | 1, 2 | 456 | |||
BT007317.1 | - | 1, 2 | 155 | |||
CR542240.1 | - | 1, 2 | 155 | |||
L36056.1 | - | 1, 2 | 155 |