Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030666.3 | NP_109591.1 | 7 | 1268 | ||
NR_073111.1 | 8 | 1400 | ||||
NR_073112.1 | 7 | 1264 | ||||
XM_011514333.1 | XP_011512635.1 | 8 | 1491 | |||
XM_011514334.2 | XP_011512636.1 | 7 | 1402 | |||
XM_011514335.2 | XP_011512637.1 | 7 | 1257 | |||
GenBank mRNA | AK223370.1 | - | 7 | 1081 | ||
AK291327.1 | - | 7 | 1081 | |||
AK298044.1 | - | 8 | 1210 | |||
AK304641.1 | - | 7 | 1113 | |||
AK312973.1 | - | 7 | 1079 | |||
BC009015.1 | - | 7 | 1066 | 3456154 | ||
BM785367.1 | - | 1 | 47 | |||
BT006928.1 | - | 6 | 1022 | |||
EU794624.1 | - | 6 | 1022 | |||
GQ472232.1 | - | 7 | 1066 | |||
M93056.1 | - | 7 | 1070 |