Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030666.3 | NP_109591.1 | 7 | 1172 | ||
NR_073111.1 | 8 | 1304 | ||||
NR_073112.1 | 7 | 1168 | ||||
XM_011514333.1 | XP_011512635.1 | 8 | 1395 | |||
XM_011514334.2 | XP_011512636.1 | 7 | 1306 | |||
XM_011514335.2 | XP_011512637.1 | 7 | 1161 | |||
GenBank mRNA | AK223370.1 | - | 7 | 985 | ||
AK291327.1 | - | 7 | 985 | |||
AK298044.1 | - | 8 | 1114 | |||
AK304641.1 | - | 7 | 1017 | |||
AK312973.1 | - | 7 | 983 | |||
BC009015.1 | - | 7 | 970 | 3456154 | ||
BI521531.1 | - | 5 | 640 | |||
BT006928.1 | - | 6 | 926 | |||
EU794624.1 | - | 6 | 926 | |||
GQ472232.1 | - | 7 | 970 | |||
M93056.1 | - | 7 | 974 |