Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELN SIRNA Assays ›
Gene Symbol:
ELN
Gene Name:
elastin
Gene Aliases:
SVAS, WBS, WS
Chromosome Location:
Chr.7: 74027772 - 74069907 on Build GRCh38
UniGene ID:
Hs.647061
Species:
Human
siRNA ID
HSS103205
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000501.3
NP_000492.2
20
1574
NM_001081752.2
NP_001075221.1
19
1544
NM_001081753.2
NP_001075222.1
20
1589
NM_001081754.2
NP_001075223.1
20
1589
NM_001081755.2
NP_001075224.1
20
1574
NM_001278912.1
NP_001265841.1
20
1574
NM_001278913.1
NP_001265842.1
17
1466
NM_001278914.1
NP_001265843.1
19
1559
NM_001278915.1
NP_001265844.1
20
1574
NM_001278916.1
NP_001265845.1
19
1532
NM_001278917.1
NP_001265846.1
19
1544
NM_001278918.1
NP_001265847.1
17
1442
NM_001278939.1
NP_001265868.1
20
1574
XM_005250187.1
XP_005250244.1
19
1215
XM_005250188.1
XP_005250245.1
19
1207
XM_011515868.1
XP_011514170.1
20
1275
XM_011515869.1
XP_011514171.1
19
1204
XM_011515870.1
XP_011514172.1
19
1570
XM_011515871.1
XP_011514173.1
19
1233
XM_011515872.1
XP_011514174.1
19
1221
XM_011515873.1
XP_011514175.1
20
1275
XM_011515874.1
XP_011514176.1
18
1540
XM_011515875.1
XP_011514177.1
18
1196
XM_011515876.1
XP_011514178.1
20
1275
XM_011515877.1
XP_011514179.1
19
1221
XM_017011813.1
XP_016867302.1
18
1118
XM_017011814.1
XP_016867303.1
19
1233
GenBank mRNA
AB208942.1
-
19
1170
AK075494.1
-
8
630
AK075554.1
-
17
1149
AK092232.1
-
17
1057
AK094138.1
-
19
1097
AK095990.1
-
20
1209
AK122731.1
-
14
837
AK125511.1
-
19
1219
AK130894.1
-
12
750
AK225659.1
-
20
1188
AK304845.1
-
20
1574
BC035570.1
-
20
1179
BC065566.1
-
19
1163
6153564
BX537939.1
-
20
1209
BX538199.1
-
19
1176
M24782.1
-
11
686
M36860.1
-
20
1224
X52896.1
-
20
1223
Back To Top
More Information
Panther Classification:
Molecular Function -
structural protein
Gene Ontology Categories:
Function(s)
Process(es)
respiratory gaseous exchange
blood circulation
cell proliferation
organ morphogenesis
extracellular matrix disassembly
extracellular matrix organization
extracellular matrix structural constituent
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline