Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001423.2 | NP_001414.1 | 5 | 593 | ||
GenBank mRNA | AK308505.1 | - | 3 | 1127 | ||
AK310798.1 | - | 3 | 321 | |||
AK312246.1 | - | 5 | 467 | |||
BC047300.1 | - | 5 | 518 | 4444217 | ||
U43916.1 | - | 4 | 346 | |||
U77085.1 | - | 5 | 562 | |||
Y07909.1 | - | 5 | 564 | |||
Z50751.1 | - | 5 | 468 |