Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L1 SIRNA Assays ›
Gene Symbol:
EPB41L1
Gene Name:
erythrocyte membrane protein band 4.1 like 1
Gene Aliases:
4.1N, MRD11
Chromosome Location:
Chr.20: 36064891 - 36232799 on Build GRCh38
UniGene ID:
Hs.437422
Species:
Human
siRNA ID
HSS103268
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258329.1
NP_001245258.1
6
695
NM_001258330.1
NP_001245259.1
6
557
NM_001258331.1
NP_001245260.1
5
457
NM_012156.2
NP_036288.2
5
641
NM_177996.2
NP_818932.1
5
452
XM_011528667.1
XP_011526969.1
10
1587
XM_011528671.1
XP_011526973.1
6
674
XM_011528673.1
XP_011526975.1
5
634
XM_011528677.1
XP_011526979.1
5
448
XM_011528681.1
XP_011526983.1
4
484
XM_011528682.1
XP_011526984.1
4
484
XM_011528683.1
XP_011526985.1
4
484
XM_011528685.1
XP_011526987.1
4
484
XM_017027711.1
XP_016883200.1
4
397
XM_017027712.1
XP_016883201.1
5
535
XM_017027713.1
XP_016883202.1
5
514
XM_017027714.1
XP_016883203.1
5
511
XM_017027715.1
XP_016883204.1
4
484
XM_017027716.1
XP_016883205.1
6
674
XM_017027717.1
XP_016883206.1
5
632
XM_017027718.1
XP_016883207.1
5
511
XM_017027719.1
XP_016883208.1
5
486
XM_017027720.1
XP_016883209.1
7
745
GenBank mRNA
AB002336.1
-
5
631
AK096848.1
-
5
504
AK126875.1
-
6
555
AK299817.1
-
5
588
AY049789.1
-
4
479
BC013885.2
-
5
457
3688606
BC040259.1
-
6
673
5195986
BI457725.1
-
4
454
CR936825.1
-
6
698
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cortical actin cytoskeleton organization
cell-cell adhesion
actin binding
structural molecule activity
protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline