Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000119.2 | NP_000110.2 | 11 | 2102 | ||
NM_001114134.1 | NP_001107606.1 | 11 | 2012 | |||
XM_005254225.1 | XP_005254282.1 | 10 | 1744 | |||
XM_011521349.2 | XP_011519651.1 | 12 | 2830 | |||
XM_011521350.2 | XP_011519652.1 | 13 | 2562 | |||
XM_011521351.2 | XP_011519653.1 | 13 | 2424 | |||
XM_011521352.2 | XP_011519654.1 | 11 | 2005 | |||
XM_011521353.2 | XP_011519655.1 | 10 | 1935 | |||
XM_011521354.2 | XP_011519656.1 | 10 | 1416 | |||
GenBank mRNA | AK297146.1 | - | 11 | 1786 | ||
AK297663.1 | - | 10 | 1662 | |||
BC096093.4 | - | 11 | 1791 | 40001236 | ||
BC096094.2 | - | 11 | 1791 | 40001240 | ||
BC096095.4 | - | 6 | 1226 | |||
BC099627.1 | - | 10 | 1575 | 40001239 | ||
BU741892.1 | - | 3 | 449 | |||
M29399.1 | - | 11 | 1899 | |||
M30646.1 | - | 11 | 1939 | |||
M30647.1 | - | 11 | 2029 | |||
M60298.1 | - | 11 | 2261 |