Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000119.2 | NP_000110.2 | 12, 13 | 2312 | ||
NM_001114134.1 | NP_001107606.1 | 12, 13 | 2222 | |||
XM_005254225.1 | XP_005254282.1 | 11, 12 | 1954 | |||
XM_011521349.2 | XP_011519651.1 | 13, 14 | 3040 | |||
XM_011521350.2 | XP_011519652.1 | 14, 15 | 2772 | |||
XM_011521351.2 | XP_011519653.1 | 14, 15 | 2634 | |||
XM_011521352.2 | XP_011519654.1 | 12, 13 | 2215 | |||
XM_011521353.2 | XP_011519655.1 | 11, 12 | 2145 | |||
XM_011521354.2 | XP_011519656.1 | 11, 12 | 1626 | |||
GenBank mRNA | AK297146.1 | - | 12, 13 | 1996 | ||
AK297663.1 | - | 11, 12 | 1872 | |||
BC096093.4 | - | 12, 13 | 2001 | 40001236 | ||
BC096094.2 | - | 12, 13 | 2001 | 40001240 | ||
BC096095.4 | - | 7, 8 | 1436 | |||
BC099627.1 | - | 11, 12 | 1785 | 40001239 | ||
BU741892.1 | - | 1, 2 | 239 | |||
M29399.1 | - | 12, 13 | 2109 | |||
M30646.1 | - | 12, 13 | 2149 | |||
M30647.1 | - | 12, 13 | 2239 | |||
M60298.1 | - | 12, 13 | 2471 |