Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005233.5 | NP_005224.2 | 3 | 674 | ||
NM_182644.2 | NP_872585.1 | 3 | 674 | |||
XM_005264715.2 | XP_005264772.1 | 3 | 599 | |||
XM_005264716.2 | XP_005264773.1 | 3 | 669 | |||
GenBank mRNA | AF213459.1 | - | 3 | 674 | ||
AF213460.1 | - | 3 | 536 | |||
AK291411.1 | - | 3 | 507 | |||
BC063282.1 | - | 3 | 578 | 30347891 | ||
HM437236.1 | - | 1 | 74 | |||
M83941.1 | - | 3 | 549 |