Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304536.1 | NP_001291465.1 | 11 | 2128 | ||
NM_001304537.1 | NP_001291466.1 | 9 | 1895 | |||
NM_004438.4 | NP_004429.1 | 10 | 1963 | |||
XM_005246374.2 | XP_005246431.1 | 10 | 1963 | |||
GenBank mRNA | AK290306.1 | - | 11 | 2128 | ||
AK300772.1 | - | 9 | 1833 | |||
AK312380.1 | - | 10 | 1909 | |||
BC026327.1 | - | 10 | 1888 | 4814429 | ||
BC105000.1 | - | 10 | 1889 | 8144003 | ||
BC105002.1 | - | 10 | 1889 | 8144005 | ||
L36645.1 | - | 10 | 1886 |