Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280794.2 | NP_001267723.2 | 9 | 2305 | ||
NM_001280795.2 | NP_001267724.2 | 6 | 1677 | |||
NM_004445.5 | NP_004436.4 | 7 | 1746 | |||
NR_104001.2 | 9 | 1592 | ||||
XM_006715881.2 | XP_006715944.1 | 10 | 2741 | |||
XM_011515879.1 | XP_011514181.1 | 11 | 2780 | |||
XM_011515880.1 | XP_011514182.1 | 10 | 2758 | |||
XM_011515881.2 | XP_011514183.1 | 10 | 2742 | |||
XM_011515882.2 | XP_011514184.1 | 10 | 2744 | |||
GenBank mRNA | BC056159.1 | - | 9 | 2174 | ||
BC110606.1 | - | 6 | 1671 | 40028676 | ||
BC110607.1 | - | 7 | 1740 | 40028687 | ||
D83492.1 | - | 10 | 2317 | |||
EU054308.1 | - | 5 | 1683 |