Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001980.3 | NP_001971.2 | 5 | 503 | ||
NM_194356.2 | NP_919337.1 | 5 | 503 | |||
XM_005253557.1 | XP_005253614.1 | 5 | 503 | |||
XM_017018978.1 | XP_016874467.1 | 5 | 500 | |||
XM_017018979.1 | XP_016874468.1 | 5 | 500 | |||
XM_017018980.1 | XP_016874469.1 | 5 | 496 | |||
XM_017018981.1 | XP_016874470.1 | 5 | 505 | |||
XM_017018982.1 | XP_016874471.1 | 5 | 500 | |||
XM_017018983.1 | XP_016874472.1 | 5 | 444 | |||
XM_017018984.1 | XP_016874473.1 | 3 | 270 | |||
XM_017018985.1 | XP_016874474.1 | 6 | 618 | |||
XM_017018986.1 | XP_016874475.1 | 5 | 519 | |||
XM_017018987.1 | XP_016874476.1 | 5 | 521 | |||
XM_017018988.1 | XP_016874477.1 | 5 | 503 | |||
GenBank mRNA | BC047496.1 | - | 5 | 495 | 5296500 | |
CD300217.1 | - | 2 | 180 | |||
CN418753.1 | - | 2 | 94 | |||
D14582.1 | - | 5 | 423 |