Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 10 | 1309 | ||
NM_001289936.1 | NP_001276865.1 | 11 | 1360 | |||
NM_001289937.1 | NP_001276866.1 | 7 | 1084 | |||
NM_001289938.1 | NP_001276867.1 | 10 | 1309 | |||
NM_004448.3 | NP_004439.2 | 7 | 1084 | |||
NR_110535.1 | 7 | 1147 | ||||
GenBank mRNA | AF177761.2 | - | 7 | 835 | ||
AK131568.1 | - | 5 | 1515 | |||
AK295195.1 | - | 11 | 1360 | |||
AK296908.1 | - | 7 | 899 | |||
AK313683.1 | - | 7 | 1038 | |||
BC080193.1 | - | 7 | 1045 | |||
BC110392.1 | - | 10 | 1274 | |||
BC167147.1 | - | 7 | 826 | 100013444 | ||
KJ534831.1 | - | 7 | 823 | |||
KJ534964.1 | - | 7 | 823 | |||
M11730.1 | - | 7 | 973 | |||
X03363.1 | - | 7 | 997 |