Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 30 | 4137 | ||
NM_001289936.1 | NP_001276865.1 | 31 | 4188 | |||
NM_001289937.1 | NP_001276866.1 | 26 | 3659 | |||
NM_004448.3 | NP_004439.2 | 27 | 3912 | |||
NR_110535.1 | 27 | 3975 | ||||
GenBank mRNA | AK295195.1 | - | 31 | 4188 | ||
AK300321.1 | - | 21 | 3086 | |||
AK313683.1 | - | 26 | 3613 | |||
BC080193.1 | - | 27 | 3873 | |||
BC167147.1 | - | 27 | 3654 | 100013444 | ||
KJ534831.1 | - | 27 | 3651 | |||
KJ534964.1 | - | 19 | 2488 | |||
M11730.1 | - | 27 | 3801 | |||
X03363.1 | - | 27 | 3825 |