Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000126.3 | NP_000117.1 | 7, 8 | 744 | ||
NM_001127716.1 | NP_001121188.1 | 6, 7 | 597 | |||
GenBank mRNA | AK292979.1 | - | 7, 8 | 744 | ||
AK300044.1 | - | 6, 7 | 532 | |||
BC015526.1 | - | 7, 8 | 688 | 3852346 | ||
BC029995.1 | - | 7, 8 | 733 | |||
BC095457.1 | - | 7, 8 | 669 | 30552923 | ||
BT009796.1 | - | 7, 8 | 663 | |||
BU176141.1 | - | 6, 7 | 560 | |||
J04058.1 | - | 7, 8 | 663 | |||
KU178046.1 | - | 7, 8 | 663 | |||
KU178047.1 | - | 8, 9 | 796 |