Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001440.3 | NP_001431.1 | 3 | 2265 | ||
XM_011544440.2 | XP_011542742.1 | 2 | 2041 | |||
GenBank mRNA | AB007042.1 | - | 3 | 2102 | ||
AB011091.1 | - | 3 | 2124 | |||
AF001690.1 | - | 3 | 2110 | |||
AF029231.1 | - | 1 | 1771 | |||
AF083551.1 | - | 3 | 2102 | |||
BC006363.2 | - | 1 | 1621 | 4131101 | ||
BT007353.1 | - | 1 | 1509 |