Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EZH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321079.1 | NP_001308008.1 | 18 | 2174 | ||
NM_001321081.1 | NP_001308010.1 | 19 | 2238 | |||
NM_001321082.1 | NP_001308011.1 | 18 | 2145 | |||
NM_001991.4 | NP_001982.2 | 19 | 2265 | |||
XM_005257145.2 | XP_005257202.1 | 17 | 2020 | |||
XM_011524517.2 | XP_011522819.1 | 18 | 2113 | |||
XM_017024350.1 | XP_016879839.1 | 18 | 2092 | |||
XM_017024351.1 | XP_016879840.1 | 17 | 1999 | |||
XM_017024352.1 | XP_016879841.1 | 16 | 1850 | |||
GenBank mRNA | AB002386.1 | - | 18 | 2177 | ||
AB004818.1 | - | 18 | 2180 | |||
AK295626.1 | - | 18 | 2127 | |||
AK299887.1 | - | 18 | 2080 | |||
AK304835.1 | - | 18 | 2078 | |||
AK309121.1 | - | 18 | 2085 | |||
AK310399.1 | - | 17 | 1957 | |||
AK310556.1 | - | 11 | 1309 | |||
AL833159.1 | - | 19 | 2185 | |||
BC015882.1 | - | 19 | 2199 | 3908776 | ||
BT009782.1 | - | 17 | 2077 | |||
U50315.1 | - | 18 | 2183 |