Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000506.4 | NP_000497.1 | 9 | 1132 | ||
NM_001311257.1 | NP_001298186.1 | 9 | 1084 | |||
GenBank mRNA | AJ972449.1 | - | 9 | 1093 | ||
AK222777.1 | - | 9 | 1067 | |||
AK293326.1 | - | 9 | 1006 | |||
AK303747.1 | - | 9 | 1046 | |||
AK312965.1 | - | 9 | 1097 | |||
AY344793.1 | - | 2 | 81 | |||
BC051332.1 | - | 9 | 1073 | 6283420 | ||
V00595.1 | - | 9 | 1043 |