Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040442.1 | NP_001035532.1 | 4 | 321 | ||
NM_001130958.1 | NP_001124430.1 | 5 | 401 | |||
NM_001445.2 | NP_001436.1 | 2 | 236 | |||
GenBank mRNA | AI208231.1 | - | 4 | 321 | ||
DQ132786.1 | - | 5 | 402 | |||
U19869.1 | - | 2 | 157 | |||
X90908.1 | - | 2 | 236 |