Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319039.1 | NP_001305968.1 | 2 | 472 | ||
NM_001319041.1 | NP_001305970.1 | 2 | 472 | |||
NM_001319042.1 | NP_001305971.1 | 2 | 213 | |||
NM_001446.4 | NP_001437.1 | 2 | 472 | |||
GenBank mRNA | AB208815.1 | - | 2 | 219 | ||
AI879148.1 | - | 2 | 515 | |||
AJ002962.1 | - | 2 | 228 | |||
AK307272.1 | - | 2 | 232 | |||
AK311867.1 | - | 2 | 231 | |||
AL512688.1 | - | 2 | 188 | |||
AW156936.1 | - | 3 | 556 | |||
BC012299.1 | - | 2 | 338 | 4707233 | ||
BI860090.1 | - | 2 | 446 | |||
BU736791.1 | - | 3 | 571 | |||
BX461253.2 | - | 2 | 193 | |||
CR457057.1 | - | 2 | 152 | |||
D50373.1 | - | 2 | 204 | |||
D88648.1 | - | 2 | 203 | |||
KM111186.1 | - | 2 | 213 | |||
U51338.1 | - | 2 | 207 | |||
U81235.1 | - | 2 | 167 |