Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004103.4 | NP_004094.3 | 22, 23 | 2288 | ||
NM_173174.2 | NP_775266.1 | 26, 27 | 2843 | |||
NM_173175.2 | NP_775267.1 | 21, 22 | 2190 | |||
NM_173176.2 | NP_775268.1 | 21, 22 | 2275 | |||
XM_005273447.4 | XP_005273504.1 | 21, 22 | 2203 | |||
XM_005273448.4 | XP_005273505.1 | 21, 22 | 2252 | |||
XM_011544441.2 | XP_011542743.1 | 21, 22 | 2213 | |||
XM_011544442.2 | XP_011542744.1 | 21, 22 | 2213 | |||
XM_017013214.1 | XP_016868703.1 | 22, 23 | 2278 | |||
XM_017013215.1 | XP_016868704.1 | 21, 22 | 2174 | |||
XM_017013216.1 | XP_016868705.1 | 21, 22 | 2213 | |||
GenBank mRNA | AB209085.1 | - | 21, 22 | 2185 | ||
AK128371.1 | - | 13, 14 | 1621 | |||
BC036651.2 | - | 26, 27 | 2843 | 4824966 | ||
BC042599.2 | - | 21, 22 | 2220 | 4343040 | ||
L49207.1 | - | 21, 22 | 2215 | |||
U33284.1 | - | 21, 22 | 2142 | |||
U43522.1 | - | 22, 23 | 2278 |