Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004461.2 | NP_004452.1 | 8 | 924 | ||
GenBank mRNA | AF042347.1 | - | 8 | 876 | ||
AK092093.1 | - | 8 | 891 | |||
AK130195.1 | - | 8 | 895 | |||
AK304587.1 | - | 7 | 803 | |||
BC006495.2 | - | 8 | 879 | 2823551 | ||
BC035438.1 | - | 4 | 344 | |||
BC043565.1 | - | 8 | 874 | |||
BT007198.1 | - | 8 | 876 | |||
CR456740.1 | - | 8 | 876 | |||
D84471.1 | - | 8 | 879 | |||
U07424.1 | - | 8 | 888 |