Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000507.3 | NP_000498.2 | 7 | 1238 | ||
NM_001127628.1 | NP_001121100.1 | 8 | 1137 | |||
XM_006717005.3 | XP_006717068.1 | 7 | 951 | |||
GenBank mRNA | AF073475.1 | - | 7 | 904 | ||
AK223395.1 | - | 7 | 1182 | |||
AK292199.1 | - | 8 | 1176 | |||
AK316583.1 | - | 7 | 1239 | |||
AY544122.1 | - | 7 | 994 | |||
BC012927.1 | - | 7 | 1104 | 4517633 | ||
D26054.1 | - | 7 | 913 | |||
D26055.1 | - | 7 | 913 | |||
D26056.1 | - | 7 | 913 | |||
L10320.1 | - | 7 | 1049 | |||
M19922.1 | - | 7 | 1105 |