Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000507.3 | NP_000498.2 | 1, 2 | 506 | ||
NM_001127628.1 | NP_001121100.1 | 2, 3 | 405 | |||
GenBank mRNA | AF073475.1 | - | 1, 2 | 172 | ||
AK223395.1 | - | 1, 2 | 450 | |||
AK292199.1 | - | 2, 3 | 444 | |||
AK316583.1 | - | 1, 2 | 507 | |||
AY544122.1 | - | 1, 2 | 262 | |||
BC012927.1 | - | 1, 2 | 372 | 4517633 | ||
D26054.1 | - | 1, 2 | 181 | |||
D26055.1 | - | 1, 2 | 181 | |||
D26056.1 | - | 1, 2 | 181 | |||
L10320.1 | - | 1, 2 | 317 | |||
M19922.1 | - | 1, 2 | 373 |