Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000139.4 | NP_000130.1 | 7 | 819 | ||
NM_001256916.1 | NP_001243845.1 | 6 | 684 | |||
XM_005273846.4 | XP_005273903.1 | 8 | 1134 | |||
XM_011544850.2 | XP_011543152.1 | 8 | 752 | |||
GenBank mRNA | BC074800.2 | - | 7 | 748 | 30915525 | |
BC074843.2 | - | 7 | 748 | 30915222 | ||
D10583.1 | - | 7 | 767 | |||
JF411082.1 | - | 6 | 582 |