Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079802.1 | NP_001073270.1 | 11 | 1555 | ||
NM_006731.2 | NP_006722.2 | 10 | 1463 | |||
XM_006717014.2 | XP_006717077.1 | 7 | 1075 | |||
XM_011518391.2 | XP_011516693.1 | 9 | 1311 | |||
XM_017014466.1 | XP_016869955.1 | 12 | 1703 | |||
XM_017014467.1 | XP_016869956.1 | 11 | 1962 | |||
XM_017014468.1 | XP_016869957.1 | 10 | 1845 | |||
XM_017014471.1 | XP_016869960.1 | 13 | 1801 | |||
XM_017014474.1 | XP_016869963.1 | 10 | 1497 | |||
XM_017014476.1 | XP_016869965.1 | 13 | 1821 | |||
XM_017014477.1 | XP_016869966.1 | 12 | 1686 | |||
XM_017014478.1 | XP_016869967.1 | 14 | 1888 | |||
XM_017014479.1 | XP_016869968.1 | 11 | 1585 | |||
GenBank mRNA | AB008226.1 | - | 10 | 1450 | ||
BC101808.1 | - | 9 | 1365 | 8069314 | ||
BC112038.1 | - | 9 | 1365 | 8327506 | ||
BC117699.1 | - | 9 | 1371 | 40073633 | ||
BC117700.1 | - | 11 | 1613 | 40073634 |