Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FKTN SIRNA Assays ›
Gene Symbol:
FKTN
Gene Name:
fukutin
Gene Aliases:
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
Chromosome Location:
Chr.9: 105558117 - 105655950 on Build GRCh38
UniGene ID:
Hs.55777
Species:
Human
siRNA ID
HSS103611
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FKTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079802.1
NP_001073270.1
8
1039
NM_001198963.1
NP_001185892.1
8
1039
NM_006731.2
NP_006722.2
7
947
XM_011518368.2
XP_011516670.1
9
1187
XM_011518369.2
XP_011516671.1
8
1037
XM_011518370.2
XP_011516672.1
9
2723
XM_011518371.2
XP_011516673.1
9
1222
XM_011518373.2
XP_011516675.1
9
1133
XM_011518374.2
XP_011516676.1
7
951
XM_011518375.2
XP_011516677.1
7
1266
XM_011518376.2
XP_011516678.1
7
1089
XM_011518378.2
XP_011516680.1
9
1187
XM_011518379.2
XP_011516681.1
7
833
XM_011518381.2
XP_011516683.1
8
1022
XM_011518389.2
XP_011516691.1
9
1187
XM_011518390.2
XP_011516692.1
5
628
XM_017014462.1
XP_016869951.1
10
1321
XM_017014463.1
XP_016869952.1
7
1410
XM_017014464.1
XP_016869953.1
9
1187
XM_017014465.1
XP_016869954.1
8
1037
XM_017014466.1
XP_016869955.1
9
1187
XM_017014467.1
XP_016869956.1
8
1446
XM_017014468.1
XP_016869957.1
7
1329
XM_017014469.1
XP_016869958.1
9
1187
XM_017014470.1
XP_016869959.1
9
1187
XM_017014471.1
XP_016869960.1
10
1285
XM_017014472.1
XP_016869961.1
10
1311
XM_017014473.1
XP_016869962.1
5
3189
XM_017014474.1
XP_016869963.1
7
950
XM_017014476.1
XP_016869965.1
10
1305
XM_017014477.1
XP_016869966.1
9
1170
XM_017014478.1
XP_016869967.1
11
1372
XM_017014479.1
XP_016869968.1
8
1069
GenBank mRNA
AB008226.1
-
7
934
AB609007.1
-
934
AK300840.1
-
8
1037
BC101808.1
-
6
849
8069314
BC112038.1
-
6
849
8327506
BC117699.1
-
6
855
40073633
BC117700.1
-
8
1097
40073634
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nervous system development
muscle organ development
negative regulation of cell proliferation
protein O-linked mannosylation
negative regulation of JNK cascade
regulation of protein glycosylation
transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0