Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308038.1 | NP_001294967.1 | 4, 5 | 572 | ||
NM_005246.3 | NP_005237.2 | 3, 4 | 591 | |||
XM_011543266.1 | XP_011541568.1 | 3, 4 | 444 | |||
XM_011543267.1 | XP_011541569.1 | 3, 4 | 2328 | |||
XM_011543269.1 | XP_011541571.1 | 2, 3 | 2182 | |||
XM_011543270.2 | XP_011541572.1 | 2, 3 | 359 | |||
XM_011543271.2 | XP_011541573.1 | 3, 4 | 444 | |||
XM_011543272.1 | XP_011541574.1 | 3, 4 | 444 | |||
XM_011543278.1 | XP_011541580.1 | 3, 4 | 444 | |||
XM_017009229.1 | XP_016864718.1 | 4, 5 | 591 | |||
XM_017009230.1 | XP_016864719.1 | 2, 3 | 359 | |||
XM_017009231.1 | XP_016864720.1 | 2, 3 | 2182 | |||
XM_017009234.1 | XP_016864723.1 | 3, 4 | 444 | |||
XM_017009235.1 | XP_016864724.1 | 3, 4 | 444 | |||
GenBank mRNA | AK293376.1 | - | 2, 3 | 445 | ||
AK296874.1 | - | 2, 3 | 166 | |||
AK315234.1 | - | 1, 2 | 279 | |||
AL707447.1 | - | 3, 4 | 479 | |||
BC017060.1 | - | 1, 2 | 273 | |||
BC058030.1 | - | 4, 5 | 532 | |||
J03358.1 | - | 3, 4 | 591 |