Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004463.2 | NP_004454.2 | 15 | 2968 | ||
GenBank mRNA | AK226117.1 | - | 15 | 2758 | ||
AK299635.1 | - | 13 | 1677 | |||
BC034530.1 | - | 15 | 2480 | 4938457 | ||
U11690.1 | - | 15 | 2964 |