Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004464.3 | NP_004455.2 | 1 | 578 | ||
NM_033143.2 | NP_149134.1 | 1 | 578 | |||
GenBank mRNA | AB016517.1 | - | 1 | 341 | ||
AF171928.1 | - | 1 | 491 | |||
AF355187.1 | - | 1 | 341 | |||
AF535149.1 | - | 1 | 503 | |||
AK291962.1 | - | 1 | 503 | |||
AK312065.1 | - | 1 | 527 | |||
BC074858.2 | - | 1 | 390 | 30915226 | ||
BC074859.2 | - | 1 | 390 | 30915463 | ||
BC131502.1 | - | 1 | 479 | 40082281 | ||
M37825.1 | - | 1 | 480 |