Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042729.1 | NP_001036194.1 | 4 | 456 | ||
NM_001042747.1 | NP_001036212.1 | 4 | 515 | |||
NM_005248.2 | NP_005239.1 | 4 | 472 | |||
XM_006710452.2 | XP_006710515.1 | 5 | 523 | |||
XM_011541010.1 | XP_011539312.1 | 4 | 437 | |||
XM_011541012.1 | XP_011539314.1 | 4 | 472 | |||
XM_011541013.2 | XP_011539315.1 | 4 | 472 | |||
GenBank mRNA | AK130188.1 | - | 4 | 434 | ||
AK301078.1 | - | 4 | 428 | |||
BC064382.1 | - | 4 | 385 | 5734966 | ||
DA139476.1 | - | 4 | 472 | |||
DA952771.1 | - | 4 | 411 | |||
DA960178.1 | - | 4 | 439 | |||
M19722.1 | - | 2 | 391 | |||
M27454.1 | - | 4 | 397 |