Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002014.3 | NP_002005.1 | 4 | 679 | ||
XM_011520929.2 | XP_011519231.1 | 4 | 1672 | |||
GenBank mRNA | AK313918.1 | - | 4 | 644 | ||
BC001786.1 | - | 4 | 634 | 3542330 | ||
BC002887.2 | - | 1 | 38 | |||
BC007924.2 | - | 4 | 579 | 4304462 | ||
M88279.1 | - | 4 | 580 |