Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145775.2 | NP_001139247.1 | 10 | 1326 | ||
NM_001145776.1 | NP_001139248.1 | 9 | 1213 | |||
NM_004117.3 | NP_004108.1 | 9 | 1168 | |||
GenBank mRNA | AF194172.1 | - | 9 | 1163 | ||
AK222807.1 | - | 9 | 1141 | |||
AK302704.1 | - | 5 | 614 | |||
AK303356.1 | - | 9 | 1030 | |||
AK312422.1 | - | 9 | 1213 | |||
BC042605.1 | - | 10 | 1326 | 4539929 | ||
BC111050.1 | - | 9 | 1129 | 5723416 | ||
BX647390.1 | - | 9 | 1141 | |||
U42031.1 | - | 8 | 987 | |||
U71321.1 | - | 9 | 1163 |