Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004475.2 | NP_004466.2 | 6 | 652 | ||
XM_005257950.3 | XP_005258007.1 | 6 | 683 | |||
XM_005257953.2 | XP_005258010.1 | 5 | 587 | |||
XM_017024394.1 | XP_016879883.1 | 6 | 580 | |||
XM_017024395.1 | XP_016879884.1 | 7 | 749 | |||
XM_017024396.1 | XP_016879885.1 | 6 | 586 | |||
GenBank mRNA | BC003683.1 | - | 6 | 564 | 3461759 | |
BC017292.1 | - | 5 | 474 | |||
BT019477.1 | - | 4 | 382 | |||
BT019478.1 | - | 4 | 382 | |||
CR541784.1 | - | 4 | 382 | |||
CR542265.1 | - | 4 | 382 | |||
M60922.1 | - | 6 | 508 |