Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FLT3LG SIRNA Assays ›
Gene Symbol:
FLT3LG
Gene Name:
fms related tyrosine kinase 3 ligand
Gene Aliases:
FL, FLT3L
Chromosome Location:
Chr.19: 49474172 - 49487038 on Build GRCh38
UniGene ID:
Hs.428
Species:
Human
siRNA ID
HSS103751
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FLT3LG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001204502.1
NP_001191431.1
5
377
NM_001204503.1
NP_001191432.1
4
346
NM_001278637.1
NP_001265566.1
5
405
NM_001278638.1
NP_001265567.1
5
394
NM_001459.3
NP_001450.2
5
377
XM_005258680.4
XP_005258737.3
4
367
XM_005258681.4
XP_005258738.3
4
367
XM_005258682.4
XP_005258739.3
4
368
XM_005258683.4
XP_005258740.3
4
367
XM_006723116.3
XP_006723179.2
4
368
XM_011526675.2
XP_011524977.1
5
413
XM_011526676.2
XP_011524978.1
5
378
XM_011526677.2
XP_011524979.1
4
367
XM_011526678.2
XP_011524980.1
5
413
XM_011526680.2
XP_011524982.1
4
367
XM_011526682.2
XP_011524984.1
4
384
XM_017026532.1
XP_016882021.1
4
368
XM_017026533.1
XP_016882022.1
4
303
XM_017026534.1
XP_016882023.1
4
276
XM_017026535.1
XP_016882024.1
5
400
GenBank mRNA
AI792400.1
-
5
358
AK301136.1
-
4
338
BC006331.2
-
5
367
BC011914.2
-
5
367
BC028001.1
-
5
378
5198812
BC126293.1
-
5
334
8992009
BC136464.1
-
5
334
9020453
BC144129.1
-
5
334
9052649
DB490139.2
-
5
408
U03858.1
-
4
350
U04806.1
-
5
359
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
signal transduction
positive regulation of cell proliferation
lymphocyte differentiation
regulation of myeloid dendritic cell activation
positive regulation of natural killer cell differentiation
embryonic hemopoiesis
positive regulation of myoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
homeostasis of number of cells within a tissue
positive regulation of cell proliferation in bone marrow
negative regulation of apoptotic process in bone marrow
positive regulation of osteoclast proliferation
positive regulation of myoblast fusion
receptor binding
cytokine activity
receptor tyrosine kinase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-pg4v9:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline