Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002020.4 | NP_002011.2 | 24 | 3336 | ||
NM_182925.4 | NP_891555.2 | 24 | 3336 | |||
XM_011534478.2 | XP_011532780.1 | 24 | 3714 | |||
XM_011534484.2 | XP_011532786.1 | 22 | 2894 | |||
XM_017009263.1 | XP_016864752.1 | 24 | 3720 | |||
XM_017009264.1 | XP_016864753.1 | 24 | 3720 | |||
XM_017009265.1 | XP_016864754.1 | 24 | 3721 | |||
XM_017009266.1 | XP_016864755.1 | 24 | 3715 | |||
XM_017009267.1 | XP_016864756.1 | 24 | 3715 | |||
XM_017009268.1 | XP_016864757.1 | 24 | 3279 | |||
GenBank mRNA | AB209637.1 | - | 17 | 2670 | ||
AK291679.1 | - | 24 | 3312 | |||
AK309910.1 | - | 24 | 3312 | |||
AY233382.1 | - | 24 | 3277 | |||
AY233383.1 | - | 24 | 3277 | |||
U43143.1 | - | 24 | 3287 | |||
X68203.1 | - | 24 | 3276 | |||
X69878.1 | - | 24 | 3278 |