Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282692.1 | NP_001269621.1 | 5 | 867 | ||
NM_001282693.1 | NP_001269622.1 | 6 | 861 | |||
NM_001282694.1 | NP_001269623.1 | 5 | 672 | |||
NM_002021.2 | NP_002012.1 | 6 | 852 | |||
XM_005245037.3 | XP_005245094.1 | 4 | 481 | |||
XM_005245038.3 | XP_005245095.1 | 6 | 861 | |||
XM_006711241.3 | XP_006711304.1 | 6 | 861 | |||
XM_006711242.3 | XP_006711305.1 | 6 | 861 | |||
GenBank mRNA | AK097039.1 | - | 7 | 990 | ||
AK290113.1 | - | 6 | 838 | |||
AK296198.1 | - | 5 | 592 | |||
AK315100.1 | - | 6 | 811 | |||
BC047129.1 | - | 5 | 833 | 5186727 | ||
M64082.1 | - | 6 | 800 |