Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002028.3 | NP_002019.1 | 6 | 698 | ||
GenBank mRNA | AK093298.1 | - | 3 | 293 | ||
AK225917.1 | - | 6 | 565 | |||
AK295972.1 | - | 3 | 262 | |||
AK315714.1 | - | 6 | 765 | |||
BC020232.1 | - | 6 | 548 | 4652202 | ||
BX248269.1 | - | 6 | 544 | |||
L00635.1 | - | 6 | 552 | |||
L10414.1 | - | 5 | 383 |
Gene Symbol: CHURC1-FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202558.1 | NP_001189487.1 | 7 | 699 | ||
NM_001202559.1 | NP_001189488.1 | 8 | 770 | |||
GenBank mRNA | AK296850.1 | - | 8 | 638 | ||
AK303739.1 | - | 7 | 575 |