Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOLH1 SIRNA Assays ›
Gene Symbol:
FOLH1
Gene Name:
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases:
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
Chromosome Location:
Chr.11: 49145094 - 49208706 on Build GRCh38
UniGene ID:
Hs.645352
Species:
Human
siRNA ID
HSS103790
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOLH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014986.1
NP_001014986.1
4, 5
784
NM_001193471.1
NP_001180400.1
5, 6
881
NM_001193472.1
NP_001180401.1
5, 6
881
NM_001193473.1
NP_001180402.1
3, 4
597
NM_004476.1
NP_004467.1
4, 5
784
XM_011519958.2
XP_011518260.2
5, 6
918
XM_017017432.1
XP_016872921.1
5, 6
916
XM_017017433.1
XP_016872922.1
5, 6
916
XM_017017434.1
XP_016872923.1
5, 6
582
XM_017017435.1
XP_016872924.1
5, 6
581
XM_017017436.1
XP_016872925.1
4, 5
816
XM_017017437.1
XP_016872926.1
4, 5
543
XM_017017438.1
XP_016872927.1
4, 5
816
XM_017017439.1
XP_016872928.1
5, 6
617
XM_017017440.1
XP_016872929.1
4, 5
487
XM_017017441.1
XP_016872930.1
4, 5
428
XM_017017442.1
XP_016872931.1
4, 5
554
XM_017017443.1
XP_016872932.1
4, 5
438
XM_017017444.1
XP_016872933.1
4, 5
718
XM_017017445.1
XP_016872934.1
5, 6
845
XM_017017446.1
XP_016872935.1
4, 5
816
XM_017017447.1
XP_016872936.1
4, 5
486
XM_017017448.1
XP_016872937.1
4, 5
478
XM_017017449.1
XP_016872938.1
4, 5
718
XM_017017450.1
XP_016872939.1
5, 6
845
XM_017017451.1
XP_016872940.1
4, 5
487
GenBank mRNA
AF176574.1
-
4, 5
661
AF254357.1
-
1, 2
46
AK295368.1
-
3, 4
529
AK295470.1
-
5, 6
813
AK300532.1
-
4, 5
716
AK312366.1
-
4, 5
712
AY101595.1
-
4, 5
523
BC025672.1
-
4, 5
682
5202715
BC108719.1
-
2, 3
288
DQ088979.1
-
4, 5
646
EF488811.2
-
5, 6
583
EF488812.1
-
5, 6
625
M99487.1
-
4, 5
784
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
folic acid-containing compound metabolic process
cellular amino acid biosynthetic process
C-terminal protein deglutamylation
metallocarboxypeptidase activity
peptidase activity
dipeptidase activity
metal ion binding
Ac-Asp-Glu binding
tetrahydrofolyl-poly(glutamate) polymer binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline