Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193306.1 | NP_001180235.1 | 3 | 921 | ||
NM_002029.3 | NP_002020.1 | 2 | 870 | |||
GenBank mRNA | AK314935.1 | - | 2 | 882 | ||
BC005315.1 | - | 2 | 863 | 3829614 | ||
BQ016000.1 | - | 1 | 460 | |||
BT007429.1 | - | 1 | 775 | |||
M37128.1 | - | 1 | 775 | |||
M60626.1 | - | 2 | 820 | |||
M60627.1 | - | 2 | 836 |
Set Membership: |
G-protein coupled receptor |